missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4-phénylphénol 98,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 207°C |
| Quantity | 1 g |
| Formula Weight | 185.23 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | 2-Amino-4-phenylphenol |
Sécurité et manipulation
| EINECSNumber | (4)-0032 |
| TSCA | Yes |