missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methylthiazole Hydrochloride 98.0+%, TCI America™

Catalog No. A03365G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A03365G 5 g
A033625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A03365G Supplier TCI America Supplier No. A03365G
Only null left

Chemical Identifiers

CAS 6142-15-0
Molecular Formula C4H7ClN2S
Molecular Weight (g/mol) 150.62
MDL Number MFCD00035214
InChI Key WUMMJVLSXMYDRW-UHFFFAOYSA-N
PubChem CID 80240
IUPAC Name hydrogen 4-methyl-1,3-thiazol-2-amine chloride
SMILES [H+].[Cl-].CC1=CSC(N)=N1

Specifications

Melting Point 173°C
Color White
Quantity 5 g
Formula Weight 150.62
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-methylthiazole Hydrochloride
EINECSNumber (5)-0203
RTECSNumber XJ2630000
TSCA Yes