missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methylpyridine, 98%

Catalog No. AAA1417714
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1417714 25 g
AAA1417722 100 g
AAA1417736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1417714 Supplier Thermo Scientific Chemicals Supplier No. A1417714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 695-34-1 | C6H8N2 | 108.144 g/mol

2-Amino-4-methylpyridine has been used in the synthesis of 2-amino-4-methyl­pyridinium 2-hy­droxy­benzoate. It is potent inhibitor of NOS2 (iNOS) in vitro. It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4-methylpyridine has been used in the synthesis of 2-amino-4-methyl­pyridinium 2-hy­droxy­benzoate. It is potent inhibitor of NOS2 (iNOS) in vitro. It is used as pharmaceutical intermediate.

Solubility
Soluble in water (41 mg/ml at 20°C), lower alcohols (slightly), chloroform, ethyl acetate, and DMF.

Notes
Hygroscopic. Incompatible with oxidizing agents and moisture. Store under dry inert gas. Protect from humidity and water. Store at -20°C.

Chemical Identifiers

CAS 695-34-1
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00006321
InChI Key ORLGLBZRQYOWNA-UHFFFAOYSA-N
Synonym 2-amino-4-methylpyridine, 2-amino-4-picoline, 2-pyridinamine, 4-methyl, ascensil, 4-methyl-2-pyridinamine, 4-methyl-2-pyridylamine, 4-methyl-2-aminopyridine, 4-picoline, 2-amino, w 45 raschig, aminton
PubChem CID 1533
IUPAC Name 4-methylpyridin-2-amine
SMILES CC1=CC(=NC=C1)N

Specifications

Melting Point 97°C to 101°C
Boiling Point 231°C
Flash Point 118°C (244°F)
Odor Unpleasant
Quantity 25 g
UN Number UN2811
Beilstein 107066
Sensitivity Hygroscopic
Merck Index 14,466
Solubility Information Soluble in water (41mg/ml at 20°C),lower alcohols (slightly),chloroform,ethyl acetate,and DMF.
Formula Weight 108.14
Percent Purity 98%
Chemical Name or Material 2-Amino-4-methylpyridine
Show More Show Less
Hazard Category H301+H311-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H318-H315-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 211-780-9
RTECSNumber TJ5150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only