missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methylbenzonitrile, 98%

Catalog No. AA4326014
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4326014 25 g
AA4326003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4326014 Supplier Thermo Scientific Chemicals Supplier No. 04326014

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 26830-96-6 | C8H8N2 | 132.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26830-96-6
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.17
MDL Number MFCD00173706
InChI Key LGNVAEIITHYWCG-UHFFFAOYSA-N
Synonym 2-cyano-5-methylaniline, 3-amino-4-cyanotoluene, 2-amino-4-methyl-benzonitrile, 4-methyl-2-aminobenzonitrile, benzonitrile, 2-amino-4-methyl, 2-amino-4-methylbenzenecarbonitrile, 2-amino-4-methylbenzonitril, 6-cyano-m-toluidine, pubchem4661, 4-methylanthranilonitrile
PubChem CID 2801276
IUPAC Name 2-amino-4-methylbenzonitrile
SMILES CC1=CC=C(C#N)C(N)=C1

Specifications

Melting Point 91°C to 93°C
Quantity 25 g
UN Number UN3439
Formula Weight 132.17
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Amino-4-methylbenzonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 248-020-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only