missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methoxybenzoic Acid 97.0+%, TCI America™

Catalog No. A23195G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23195G 5 g
A23191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23195G Supplier TCI America Supplier No. A23195G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4294-95-5
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00667729
InChI Key HHNWXQCVWVVVQZ-UHFFFAOYSA-N
Synonym 4-methoxyanthranilic acid, 2-amino-4-methoxy-benzoic acid, 2-amino-p-anisic acid, benzoic acid, 2-amino-4-methoxy, 3-amino-4-carboxyanisole, 2-carboxy-5-methoxyaniline, 4-methoxyanthranilicacid, 2-amine-4-methoxybenzoic acid, pubchem11856, acmc-1aerh
PubChem CID 351010
IUPAC Name 2-amino-4-methoxybenzoic acid
SMILES COC1=CC(=C(C=C1)C(=O)O)N

Specifications

Melting Point 167°C
Quantity 5 g
Formula Weight 167.16
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-methoxybenzoic Acid