missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-fluorobenzoic acid, 96%

Catalog No. AC293020025
Change view
Click to view available options
Quantity:
2.5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC293020025 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC293020025 Supplier Thermo Scientific Chemicals Supplier No. 293020025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 446-32-2 | C7H6FNO2 | 155.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 446-32-2
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.13
MDL Number MFCD00075553
InChI Key LGPVTNAJFDUWLF-UHFFFAOYSA-N
Synonym 4-fluoroanthranilic acid, 2-amino-4-fluoro-benzoic acid, benzoic acid, 2-amino-4-fluoro, 2-amino-4-fluoro benzoic acid, 2-amino-4-fluorobenzoicacid, fa0, pubchem1323, amino-4-fluorobenzoic acid, acmc-209jz7, 2-amino-4fluorobenzoic acid
PubChem CID 2724967
IUPAC Name 2-amino-4-fluorobenzoic acid
SMILES C1=CC(=C(C=C1F)N)C(=O)O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 188.0°C to 196.0°C
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Linear Formula H2NC6H3(F)COOH
Quantity 2.5 g
Formula Weight 155.13
Percent Purity 96%
Chemical Name or Material 2-Amino-4-fluorobenzoic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-163-9

RUO – Research Use Only