missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chloropyridine, 97%

Catalog No. AAH5913014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5913014 25 g
AAH5913006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5913014 Supplier Thermo Scientific Chemicals Supplier No. H5913014
Only null left

CAS: 19798-80-2 | C5H6ClN2 | 129.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19798-80-2
Molecular Formula C5H6ClN2
Molecular Weight (g/mol) 129.57
MDL Number MFCD04113820
InChI Key RQMWVVBHJMUJNZ-UHFFFAOYSA-O
Synonym 2-amino-4-chloropyridine, 4-chloro-pyridin-2-ylamine, 2-pyridinamine, 4-chloro, 4-chloro-2-pyridinamine, 4-chloro-2-pyridylamine, 2-amino-4-chloro pyridine, 4-chloro-2-pyridinylamine, 2-amino-4-chloro-pyridine, 4-chloro-2-aminopyridine, 4-chlorpyridin-2-amin
PubChem CID 581866
IUPAC Name 4-chloropyridin-2-amine
SMILES NC1=CC(Cl)=CC=[NH+]1

Specifications

Melting Point 129°C to 133°C
Quantity 25 g
Sensitivity Air and moisture sensitive
Formula Weight 128.56
Percent Purity 97%
Chemical Name or Material 2-Amino-4-chloropyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only