missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chlorobenzenethiol 98.0+%, TCI America™

Catalog No. A16255G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16255G 5 g
A162525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A16255G Supplier TCI America Supplier No. A16255G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1004-00-8
Molecular Formula C6H6ClNS
Molecular Weight (g/mol) 159.631
MDL Number MFCD00792528
InChI Key NGIRMPARLVGMPX-UHFFFAOYSA-N
Synonym 2-amino-4-chlorothiophenol, 2-amino-4-chlorobenzene-1-thiol, 2-amino-4-chloro-benzenethiol, benzenethiol, 2-amino-4-chloro, 4-chloro-2-aminothiophenol, 5-chloro-2-mercaptoaniline, 2-amino-4-chloro thiophenol, 5-chloro-2-mercaptoaniline hydrochloride, benzenethiol, 2-amino-4-chloro-, hydrochloride, pubchem5427
PubChem CID 12000
IUPAC Name 2-amino-4-chlorobenzenethiol
SMILES C1=CC(=C(C=C1Cl)N)S

Specifications

Melting Point 58°C
Quantity 5 g
UN Number 3259
Formula Weight 159.63
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-chlorobenzenethiol