missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,6-dihydroxypyrimidine 98.0+%, TCI America™

Catalog No. A109925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A109925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A109925G Supplier TCI America Supplier No. A109925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 56-09-7
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.10
MDL Number MFCD00006094
InChI Key AUFJTVGCSJNQIF-UHFFFAOYSA-N
Synonym 2-amino-4,6-dihydroxypyrimidine, 2-aminopyrimidine-4,6-diol, 4 1h-pyrimidinone, 2-amino-6-hydroxy, 2-amino-6-hydroxypyrimidin-4 3h-one, 2-amino-4,6-dioxypyrimidine, 2-amino-4,6-pyrimidinedione, 4,6-dihydroxy-2-aminopyrimidine, 2-amino-4,6-pyrimidinediol, 2-amino-6-hydroxy-1h-pyrimidin-4-one, 2-amino-6-hydroxy-4 1h-pyrimidinone
PubChem CID 66131
IUPAC Name 2-amino-6-hydroxy-3,4-dihydropyrimidin-4-one
SMILES NC1=NC(O)=CC(=O)N1

Specifications

Quantity 25 g
Formula Weight 127.10
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4,6-dihydroxypyrimidine
RTECSNumber UW7361000
TSCA Yes
Recommended Storage Refrigerator