missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4,5-dimethylbenzoic Acid 97.0+%, TCI America™
Supplier: TCI America A28501G
Specifications
| 2-Amino-4,5-dimethylbenzoic Acid | |
| 208°C | |
| C9H11NO2 | |
| 1 g | |
| REBBDMHTSKPGCO-UHFFFAOYSA-N | |
| 2-amino-4,5-dimethylbenzoic acid | |
| 266534 | |
| ≥97.0% (HPLC,T) |
| 15089-51-7 | |
| Yellow | |
| MFCD00156998 | |
| 4,5-Dimethylanthranilic Acid | |
| CC1=C(C=C(C(=C1)C(=O)O)N)C | |
| 165.192 | |
| 165.19 | |
| Crystalline Powder |
Chemical Identifiers
| 15089-51-7 | |
| 165.192 | |
| REBBDMHTSKPGCO-UHFFFAOYSA-N | |
| 266534 | |
| CC1=C(C=C(C(=C1)C(=O)O)N)C |
| C9H11NO2 | |
| MFCD00156998 | |
| 4,5-Dimethylanthranilic Acid | |
| 2-amino-4,5-dimethylbenzoic acid |
Safety and Handling
TSCA : No