CAS RN 15089-51-7
IUPAC Name:
2-amino-4,5-dimethylbenzoic acid
Synonyms:
2-amino-4,5-dimethylbenzoic acid
Molecular Weight (g/mol):
165.19
Molecular Formula:
C9H11NO2
InChi Key:
REBBDMHTSKPGCO-UHFFFAOYSA-N
SMILES:
CC1=CC(N)=C(C=C1C)C(O)=O
1
–
1
of
1
results
1
2-Amino-4,5-dimethylbenzoic Acid 97.0+%, TCI America™
CAS: 15089-51-7 Molecular Formula: C9H11NO2 Molecular Weight (g/mol): 165.192 MDL Number: MFCD00156998 InChI Key: REBBDMHTSKPGCO-UHFFFAOYSA-N Synonym: 4,5-Dimethylanthranilic Acid PubChem CID: 266534 IUPAC Name: 2-amino-4,5-dimethylbenzoic acid SMILES: CC1=C(C=C(C(=C1)C(=O)O)N)C