missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,5-dimethylbenzoic Acid 97.0+%, TCI America™

Catalog No. p-7118110
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2850200MG 200 mg
A28501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2850200MG Supplier TCI America Supplier No. A2850200MG
Only null left

Chemical Identifiers

CAS 15089-51-7
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00156998
InChI Key REBBDMHTSKPGCO-UHFFFAOYSA-N
Synonym 4,5-Dimethylanthranilic Acid
PubChem CID 266534
IUPAC Name 2-amino-4,5-dimethylbenzoic acid
SMILES CC1=C(C=C(C(=C1)C(=O)O)N)C

Specifications

Melting Point 208°C
Color Yellow
Quantity 200 mg
Formula Weight 165.19
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4,5-dimethylbenzoic Acid