missing translation for 'onlineSavingsMsg'
Learn More

2'-Amino-4',5'-dimethoxyacetophenone 98.0+%, TCI America™

Catalog No. A28421G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28421G 1 g
A28425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28421G Supplier TCI America Supplier No. A28421G

Chemical Identifiers

CAS 4101-30-8
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.218
MDL Number MFCD00016646
InChI Key KGKWXEGYKGTMAK-UHFFFAOYSA-N
Synonym 1-2-amino-4,5-dimethoxyphenyl ethanone, 2'-amino-4',5'-dimethoxyacetophenone, 2-amino-4,5-dimethoxyacetophenone, 1-2-amino-4,5-dimethoxyphenyl ethan-1-one, 1-2-amino-4,5-dimethoxyhenyl ethanone, ethanone, 1-2-amino-4,5-dimethoxyphenyl, 1-acetyl-2-amino-4,5-dimethoxybenzene, pubchem4007, acmc-1arib, 2-acetyl-4,5-dimethoxyaniline
PubChem CID 602085
IUPAC Name 1-(2-amino-4,5-dimethoxyphenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1N)OC)OC

Specifications

Melting Point 107°C
Color Yellow
Quantity 1 g
Formula Weight 195.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Amino-4′,5′-dimethoxyacetophenone