missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-nitrophenol, 98%

Catalog No. AAH2747214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2747214 25 g
AAH2747206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2747214 Supplier Thermo Scientific Chemicals Supplier No. H2747214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 603-85-0 | C6H6N2O3 | 154.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 603-85-0
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.13
MDL Number MFCD00010875
InChI Key KUCWUAFNGCMZDB-UHFFFAOYSA-N
Synonym phenol, 2-amino-3-nitro, 2-hydroxy-6-nitroaniline, ccris 2544, unii-a7f97zpu2w, 3-nitro-2-aminophenol, 2-amino-3-nitro-phenol, a7f97zpu2w, 2-amino-3-hydroxynitrobenzene, 1-hydroxy-2-amino-3-nitrobenzene, 2-amino-nitrophenol
PubChem CID 4115495
IUPAC Name 2-amino-3-nitrophenol
SMILES NC1=C(O)C=CC=C1[N+]([O-])=O

Specifications

Melting Point ∼212°C
Quantity 25 g
Beilstein 2804307
Formula Weight 154.13
Percent Purity 98%
Chemical Name or Material 2-Amino-3-nitrophenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-060-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only