missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-methoxybenzoic Acid 99.0+%, TCI America™

Supplier:  TCI America A137825G

 View more versions of this product

Catalog No. A137825G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Amino-3-methoxybenzoic Acid
172°C
MFCD00075178
3-methoxyanthranilic acid, benzoic acid, 2-amino-3-methoxy, 3-methoxyanthranilate, m-anisic acid, 2-amino, 3-methoxy-2-aminobenzoic acid, 2-amino-3-methoxy-benzoic acid, 2-amino-3-methoxy benzoic acid, 2-amino-3-methyloxy benzoic acid, 2-amino-3-methoxybenzoicacid
COC1=CC=CC(=C1N)C(=O)O
167.164
CHEBI:27440
≥99.0% (HPLC,T)
3177-80-8
C8H9NO3
25 g
SXOPCLUOUFQBJV-UHFFFAOYSA-N
2-amino-3-methoxybenzoic acid
255720
167.16
Crystalline Powder

Chemical Identifiers

3177-80-8
167.164
SXOPCLUOUFQBJV-UHFFFAOYSA-N
255720
2-amino-3-methoxybenzoic acid
C8H9NO3
MFCD00075178
3-methoxyanthranilic acid, benzoic acid, 2-amino-3-methoxy, 3-methoxyanthranilate, m-anisic acid, 2-amino, 3-methoxy-2-aminobenzoic acid, 2-amino-3-methoxy-benzoic acid, 2-amino-3-methoxy benzoic acid, 2-amino-3-methyloxy benzoic acid, 2-amino-3-methoxybenzoicacid
CHEBI:27440
COC1=CC=CC(=C1N)C(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions