missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-methoxybenzoic Acid 99.0+%, TCI America™

Catalog No. p-7117399
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13785G 5 g
A137825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A13785G Supplier TCI America Supplier No. A13785G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3177-80-8
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00075178
InChI Key SXOPCLUOUFQBJV-UHFFFAOYSA-N
Synonym 3-methoxyanthranilic acid, benzoic acid, 2-amino-3-methoxy, 3-methoxyanthranilate, m-anisic acid, 2-amino, 3-methoxy-2-aminobenzoic acid, 2-amino-3-methoxy-benzoic acid, 2-amino-3-methoxy benzoic acid, 2-amino-3-methyloxy benzoic acid, 2-amino-3-methoxybenzoicacid
PubChem CID 255720
ChEBI CHEBI:27440
IUPAC Name 2-amino-3-methoxybenzoic acid
SMILES COC1=CC=CC(=C1N)C(=O)O

Specifications

Melting Point 172°C
Quantity 5 g
Formula Weight 167.16
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-methoxybenzoic Acid