accessibility menu, dialog, popup

UserName

CH3OOHOH2N

CAS RN 3177-80-8

IUPAC Name: 2-amino-3-methoxybenzoic acid
Synonyms: 2-amino-3-methoxybenzoic acid
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: SXOPCLUOUFQBJV-UHFFFAOYSA-N
SMILES: COC1=CC=CC(C(O)=O)=C1N
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)