missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-bromo-5-chloropyridine 98.0+%, TCI America™

Catalog No. A24025G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24025G 5 g
A240225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24025G Supplier TCI America Supplier No. A24025G
Only null left

Chemical Identifiers

CAS 26163-03-1
Molecular Formula C5H4BrClN2
Molecular Weight (g/mol) 207.455
MDL Number MFCD00955627
InChI Key UWGGGYYCKDCTGN-UHFFFAOYSA-N
Synonym 2-amino-3-bromo-5-chloropyridine, 3-bromo-5-chloro-2-pyridinamine, 2-pyridinamine, 3-bromo-5-chloro, 3-bromo-5-chloro-2-pyridylamine, 3-bromo-5-chloropyrid-2-ylamine, 3-bromo-5-chloro-pyridin-2-ylamine, abbypharma ap-10-5406, 3-bromo-5-chloro-pyridin-2-amine, pubchem1141, acmc-1ch3b
PubChem CID 2763971
IUPAC Name 3-bromo-5-chloropyridin-2-amine
SMILES C1=C(C=NC(=C1Br)N)Cl

Specifications

Melting Point 84°C
Quantity 5 g
Formula Weight 207.46
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-bromo-5-chloropyridine