missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,5-dichloropyridine, 98%

Catalog No. AAB2198422
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2198422 100 g
AAB2198414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2198422 Supplier Thermo Scientific Chemicals Supplier No. B2198422

CAS: 4214-74-8 | C5H4Cl2N2 | 163.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4214-74-8
Molecular Formula C5H4Cl2N2
Molecular Weight (g/mol) 163.00
MDL Number MFCD00006313
InChI Key OCWBGKZFOYMCCN-UHFFFAOYSA-N
Synonym 2-amino-3,5-dichloropyridine, 3,5-dichloro-2-pyridinamine, 2-pyridinamine, 3,5-dichloro, 3,5-dichloro-3-pyridylamine, 3,5-dichloro-2-aminopyridine, 3,5-dichloro-2-pyridylamine, 3,5-dichloropyridin-2-ylamine, pyridine, 2-amino-3,5-dichloro, pubchem13504, acmc-209jnv
PubChem CID 77886
IUPAC Name 3,5-dichloropyridin-2-amine
SMILES NC1=NC=C(Cl)C=C1Cl

Specifications

Melting Point 81°C to 83°C
Odor Unpleasant
Quantity 100 g
Beilstein 119376
Formula Weight 163.01
Percent Purity 98%
Chemical Name or Material 2-Amino-3,5-dichloropyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 224-143-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only