missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,5-dibromopyridine, 97%

Catalog No. AAA1270622
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1270622 100 g
AAA1270614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1270622 Supplier Thermo Scientific Chemicals Supplier No. A1270622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 35486-42-1 | C5H4Br2N2 | 251.909 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35486-42-1
Molecular Formula C5H4Br2N2
Molecular Weight (g/mol) 251.909
MDL Number MFCD00038041
InChI Key WJMJWMSWJSACSN-UHFFFAOYSA-N
Synonym 2-amino-3,5-dibromopyridine, 3,5-dibromo-2-pyridylamine, 2-pyridinamine, 3,5-dibromo, 3,5-dibromo-2-pyridinamine, 3,5-dibromo-pyridin-2-ylamine, 3,5-dibromo-2-pyridinylamine, 2-amino-3,5-dibromo pyridine, 3,5-dibrompyridin-2-amin, pubchem1130, maybridge1_001693
PubChem CID 98851
IUPAC Name 3,5-dibromopyridin-2-amine
SMILES C1=C(C=NC(=C1Br)N)Br

Specifications

Melting Point 102°C to 105°C
Quantity 100 g
Beilstein 119390
Formula Weight 251.92
Percent Purity 97%
Chemical Name or Material 2-Amino-3,5-dibromopyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 252-590-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only