missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,5-dibromobenzoic acid, Thermo Scientific™

Catalog No. BTB01924EA
Change view
Click to view available options
Quantity:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB01924EA 10g
BTB01924EB 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB01924EA Supplier Thermo Scientific Chemicals Supplier No. BTB01924EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 609-85-8
Molecular Formula C7H5Br2NO2
Molecular Weight (g/mol) 294.93
InChI Key WNABMWFLKQEGCP-UHFFFAOYSA-N
Synonym 3,5-dibromoanthranilic acid, 2-amino-3,5-dibromobenzoicacid, 3,5-dibromo-2-aminobenzoic acid, buttpark 89\07-50, 2-amino-3,5-dibromo-benzoic acid, benzoic acid, 2-amino-3,5-dibromo, acmc-20ahts, pubchem3720, 2-carboxy-4,6-dibromoaniline, ghl.pd_mitscher_leg0.1128
PubChem CID 219491
IUPAC Name 2-amino-3,5-dibromobenzoic acid
SMILES C1=C(C=C(C(=C1Br)N)C(=O)O)Br

Specifications

CAS Min % 80.0
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 10g
Formula Weight 294.93
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-Amino-3, 5-dibromobenzoic acid
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.