missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-3,5-dibromo-6-methylpyridine 98.0+%, TCI America™
Supplier: TCI America A16631G
Specifications
| 2-Amino-3,5-dibromo-6-methylpyridine | |
| 134°C | |
| MFCD00068229 | |
| 2-amino-3,5-dibromo-6-methylpyridine, 3,5-dibromo-6-methyl-2-pyridinamine, 2-amino-3,5-dibromo-6-picoline, 6-amino-3,5-dibromo-2-picoline, 3,5-dibromo-6-methyl-pyridin-2-ylamine, 3,5-dibromo-6-methyl-2-pyridylamine, 2-pyridinamine, 3,5-dibromo-6-methyl, wisteria sinensis extract, 2-amino-3,5-dibrom-6-methylpyridine, pubchem7669 | |
| CC1=NC(=C(C=C1Br)Br)N | |
| 265.936 | |
| 265.94 | |
| Crystalline Powder |
| 91872-10-5 | |
| C6H6Br2N2 | |
| 1 g | |
| BLUYMDMBXIMIGK-UHFFFAOYSA-N | |
| 3,5-dibromo-6-methylpyridin-2-amine | |
| 522994 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 91872-10-5 | |
| 265.936 | |
| BLUYMDMBXIMIGK-UHFFFAOYSA-N | |
| 522994 | |
| CC1=NC(=C(C=C1Br)Br)N |
| C6H6Br2N2 | |
| MFCD00068229 | |
| 2-amino-3,5-dibromo-6-methylpyridine, 3,5-dibromo-6-methyl-2-pyridinamine, 2-amino-3,5-dibromo-6-picoline, 6-amino-3,5-dibromo-2-picoline, 3,5-dibromo-6-methyl-pyridin-2-ylamine, 3,5-dibromo-6-methyl-2-pyridylamine, 2-pyridinamine, 3,5-dibromo-6-methyl, wisteria sinensis extract, 2-amino-3,5-dibrom-6-methylpyridine, pubchem7669 | |
| 3,5-dibromo-6-methylpyridin-2-amine |
Safety and Handling
TSCA : No