missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Amino-2'-deoxyguanosine, 98%

Catalog No. AAJ64650MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64650MD 250 mg
AAJ6465003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ64650MD Supplier Thermo Scientific Chemicals Supplier No. J64650MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 60966-26-9 | C10H14N6O4 | 282.26 g/mol

2‘-Amino-2‘-deoxyguanosine is a valuable probe for investigating the metal ion interactions at the active site of the group I ribozyme.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2‘-Amino-2‘-deoxyguanosine is a valuable probe for investigating the metal ion interactions at the active site of the group I ribozyme.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Chemical Identifiers

CAS 60966-26-9
Molecular Formula C10H14N6O4
Molecular Weight (g/mol) 282.26
MDL Number MFCD01723954
InChI Key ROPTVRLUGSPXNH-SJLGBTOWNA-N
PubChem CID 173775
IUPAC Name 2-amino-9-[(2R,3R,4S,5R)-3-amino-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
SMILES N[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C1N=C(N)NC2=O

Specifications

Color White
Quantity 250 mg
Synonym 2'-amino-2'-deoxyguanosine, guanosine, 2'-amino-2'-deoxy, 2'-deoxy-2'-aminoguanosine, 2ag, 2-amino-9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl oxolan-2-yl-1h-purin-6-one, guanosine,2'-amino-2'-deoxy, 2'-amino-2'-deoxyguanosine,2'-nh2-dg, 2-amino-9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-purin-6-one, 2-amino-9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1,9-dihydro-6h-purin-6-one, 2-amino-9-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6-one
Formula Weight 282.26
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2'-Amino-2'-deoxyguanosine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
RTECSNumber MF8752000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only