missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-1-pyrroline Hydrochloride 98.0+%, TCI America™

Catalog No. A29631G
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
Catalog No. Quantity
A29631G 1 g
1 options

Catalog No. A29631G

Supplier: TCI America A29631G

Only null left

Chemical Identifiers

CAS 7544-75-4
Molecular Formula C4H9ClN2
Molecular Weight (g/mol) 120.58
MDL Number MFCD00790772
InChI Key JEBHLIYFPKISBI-UHFFFAOYSA-N
Synonym 3,4-Dihydro-2H-pyrrol-5-amine Hydrochloride
PubChem CID 12778905
IUPAC Name 3,4-dihydro-2H-pyrrol-5-amine hydrochloride
SMILES Cl.NC1=NCCC1

Specifications

Melting Point 173°C
Color White
Quantity 1 g
Formula Weight 120.58
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-1-pyrroline Hydrochloride