missing translation for 'onlineSavingsMsg'
Learn More

2-Allylaniline Hydrochloride 98.0+%, TCI America™

Catalog No. A29945G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A29945G Supplier TCI America Supplier No. A29945G
Only null left

Chemical Identifiers

CAS 138286-02-9
Molecular Formula C9H12ClN
Molecular Weight (g/mol) 169.652
InChI Key QEUTZGMYXDQKNU-UHFFFAOYSA-N
PubChem CID 85622391
IUPAC Name 2-prop-2-enylaniline;hydrochloride
SMILES C=CCC1=CC=CC=C1N.Cl

Specifications

Melting Point 132°C
Color White
Quantity 5 g
Formula Weight 169.65
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Allylaniline Hydrochloride