Learn More
2-Allyl-N-Fmoc-L-glycine, 95%
CAS: 146549-21-5 | C20H19NO4 | 337.375 g/mol
$99.90 - $1135.10
Chemical Identifiers
| CAS | 146549-21-5 |
|---|---|
| Molecular Formula | C20H19NO4 |
| Molecular Weight (g/mol) | 337.375 |
| MDL Number | MFCD01311749 |
| InChI Key | YVBLQCANYSFEBN-SFHVURJKSA-N |
| Synonym | fmoc-l-allylglycine, fmoc-gly allyl-oh, fmoc-l-allyglycine, s-2-9h-fluoren-9-yl methoxy carbonyl amino pent-4-enoic acid, fmoc-allyl-gly-oh, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino pent-4-enoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino pent-4-enoic acid, 4-pentenoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-, 2s, s-n-fmoc-allyl-glycine, s-n-fmoc-allylglycine |
| PubChem CID | 2734457 |
| IUPAC Name | (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pent-4-enoic acid |
| SMILES | C=CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH52177MD
|
Thermo Scientific Chemicals
H52177MD |
250 mg |
Each for $99.90
|
|
|||||
|
AAH5217703
|
Thermo Scientific Chemicals
H5217703 |
1 g |
Each for $280.68
|
|
|||||
|
AAH5217706
|
Thermo Scientific Chemicals
H5217706 |
5 g |
Each for $1,135.10
|
|
|||||
Description
2-Allyl-N-Fmoc-L-glycine is potentially useful for solid phase peptide synthesis techniques. 2-Allyl-N-Fmoc-L-glycine compound could be useful as an unusual amino acid analog to aid in the deconvolution of protein structure and function. Reagent in the synthesis of a Dicarba-Analog of Octreotide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Allyl-N-Fmoc-L-glycine is potentially useful for solid phase peptide synthesis techniques. 2-Allyl-N-Fmoc-L-glycine compound could be useful as an unusual amino acid analog to aid in the deconvolution of protein structure and function. Reagent in the synthesis of a Dicarba-Analog of Octreotide.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.
Chemical Identifiers
| 146549-21-5 | |
| 337.375 | |
| YVBLQCANYSFEBN-SFHVURJKSA-N | |
| 2734457 | |
| C=CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 |
| C20H19NO4 | |
| MFCD01311749 | |
| fmoc-l-allylglycine, fmoc-gly allyl-oh, fmoc-l-allyglycine, s-2-9h-fluoren-9-yl methoxy carbonyl amino pent-4-enoic acid, fmoc-allyl-gly-oh, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino pent-4-enoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino pent-4-enoic acid, 4-pentenoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-, 2s, s-n-fmoc-allyl-glycine, s-n-fmoc-allylglycine | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pent-4-enoic acid |
Specifications
| 146549-21-5 | |
| MFCD01311749 | |
| 5884047 | |
| Slightly soluble in water. | |
| C=CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 | |
| 337.375 | |
| 337.37 | |
| 2-Allyl-N-Fmoc-L-glycine |
| C20H19NO4 | |
| 250 mg | |
| fmoc-l-allylglycine, fmoc-gly allyl-oh, fmoc-l-allyglycine, s-2-9h-fluoren-9-yl methoxy carbonyl amino pent-4-enoic acid, fmoc-allyl-gly-oh, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino pent-4-enoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino pent-4-enoic acid, 4-pentenoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-, 2s, s-n-fmoc-allyl-glycine, s-n-fmoc-allylglycine | |
| YVBLQCANYSFEBN-SFHVURJKSA-N | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pent-4-enoic acid | |
| 2734457 | |
| 95% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only