accessibility menu, dialog, popup

UserName

OO(S)H2CNHOO

CAS RN 146549-21-5

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pent-4-enoate
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}pent-4-enoate
Molecular Weight (g/mol): 336.37
Molecular Formula: C20H18NO4
InChi Key: YVBLQCANYSFEBN-SFHVURJKSA-M
SMILES: [O-]C(=O)[C@H](CC=C)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)