Learn More
2-Allyl-N-Fmoc-L-glycine, 95%
CAS: 146549-21-5 | C20H19NO4 | 337.375 g/mol
Supplier: Thermo Scientific Chemicals H5217703
Description
2-Allyl-N-Fmoc-L-glycine is potentially useful for solid phase peptide synthesis techniques. 2-Allyl-N-Fmoc-L-glycine compound could be useful as an unusual amino acid analog to aid in the deconvolution of protein structure and function. Reagent in the synthesis of a Dicarba-Analog of Octreotide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Allyl-N-Fmoc-L-glycine is potentially useful for solid phase peptide synthesis techniques. 2-Allyl-N-Fmoc-L-glycine compound could be useful as an unusual amino acid analog to aid in the deconvolution of protein structure and function. Reagent in the synthesis of a Dicarba-Analog of Octreotide.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.
Specifications
| 2-Allyl-N-Fmoc-L-glycine | |
| C20H19NO4 | |
| 1 g | |
| fmoc-l-allylglycine, fmoc-gly allyl-oh, fmoc-l-allyglycine, s-2-9h-fluoren-9-yl methoxy carbonyl amino pent-4-enoic acid, fmoc-allyl-gly-oh, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino pent-4-enoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino pent-4-enoic acid, 4-pentenoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-, 2s, s-n-fmoc-allyl-glycine, s-n-fmoc-allylglycine | |
| YVBLQCANYSFEBN-SFHVURJKSA-N | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pent-4-enoic acid | |
| 2734457 | |
| 95% |
| 146549-21-5 | |
| MFCD01311749 | |
| 5884047 | |
| Slightly soluble in water. | |
| C=CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 | |
| 337.375 | |
| 337.37 |
Chemical Identifiers
| 146549-21-5 | |
| 337.375 | |
| YVBLQCANYSFEBN-SFHVURJKSA-N | |
| 2734457 | |
| C=CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 |
| C20H19NO4 | |
| MFCD01311749 | |
| fmoc-l-allylglycine, fmoc-gly allyl-oh, fmoc-l-allyglycine, s-2-9h-fluoren-9-yl methoxy carbonyl amino pent-4-enoic acid, fmoc-allyl-gly-oh, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino pent-4-enoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino pent-4-enoic acid, 4-pentenoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-, 2s, s-n-fmoc-allyl-glycine, s-n-fmoc-allylglycine | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pent-4-enoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only