missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylpyridine, 98%

Catalog No. AC102680250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC102680250 25 g Glass bottle
AC102681000 100 g Glass bottle
AC102685000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC102680250 Supplier Thermo Scientific Chemicals Supplier No. 102680250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1122-62-9 | C7H7NO | 121.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1122-62-9
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.14
InChI Key AJKVQEKCUACUMD-UHFFFAOYSA-N
Synonym 2-acetylpyridine, 1-pyridin-2-yl ethanone, methyl 2-pyridyl ketone, 2-acetopyridine, 2-pyridyl methyl ketone, acetylpyridine, ethanone, 1-2-pyridinyl, ketone, methyl 2-pyridyl, 1-pyridin-2-yl ethan-1-one, 1-2-pyridinyl ethanone
PubChem CID 14286
IUPAC Name 1-pyridin-2-ylethanone
SMILES CC(=O)C1=CC=CC=N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 8.0°C to 10.0°C
Color Colorless to Brown
Density 1.0800g/mL
Boiling Point 188.0°C to 189.0°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5200 to 1.5220
Linear Formula C5H4NCOCH3
Quantity 25 g
Specific Gravity 1.08
Solubility Information Solubility in water: 200g/L (20°C). Other solubilities: miscible with ethanol
Formula Weight 121.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Acetylpyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 214-355-6

RUO – Research Use Only