missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-4-(1,2,3,4-tetrahydroxybutyl)imidazole, 97%

Catalog No. AAH63416MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63416MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH63416MD Supplier Thermo Scientific Chemicals Supplier No. H63416MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 94944-70-4 | C9H14N2O5 | 230.22 g/mol

2-Acetyl-4-(1,2,3,4-tetrahydroxybutyl)imidazole used for treatment of autoimmune disorders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetyl-4-(1,2,3,4-tetrahydroxybutyl)imidazole used for treatment of autoimmune disorders.

Solubility
Soluble in distilled water, DMSO (>5 mg/ml), clear (warmed), and PBS pH 7.2 (∼0.15 mg/ml).

Notes

Chemical Identifiers

CAS 94944-70-4
Molecular Formula C9H14N2O5
Molecular Weight (g/mol) 230.22
MDL Number MFCD08752516
InChI Key CQSIXFHVGKMLGQ-BWZBUEFSSA-N
Synonym 2-athbi, 2-acetyl-4 5-tetrahydroxybutylimidazole, unii-hw195j55g1, thi, 2-acetyl-5-tetrahydroxybutyl imidazole, 2-acetyl-4-tetrahydroxybutylimidazole, ethanone, 1-4-1,2,3,4-tetrahydroxybutyl-1h-imidazol-2-yl-, 1r-1r*,2s*,3r*, 2-acetyl-4 5-1,2,3,4-tetrahydroxybutyl-imidazole thi, 1-5-1r,2s,3r-1,2,3,4-tetrahydroxybutyl-1h-imidazol-2-yl ethanone, 2-acetyl-4-tetrahydroxybutyl imidazole; 1-5-1r,2s,3r-1,2,3,4-tetrahydroxybutyl-1h-imidazol-2-yl-ethanone
PubChem CID 108037
IUPAC Name 1-[5-[(1R,2S,3R)-1,2,3,4-tetrahydroxybutyl]-1H-imidazol-2-yl]ethanone
SMILES CC(=O)C1=NC=C(N1)C(C(C(CO)O)O)O

Specifications

Color White to Brown
Quantity 250 mg
Solubility Information Soluble in distilled water,DMSO (>5mg/ml),clear (warmed),and PBS pH 7.2 (∽0.15mg/ml).
Formula Weight 230.22
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Acetyl-4-(1,2,3,4-tetrahydroxybutyl)imidazole
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only