missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamido-4-methyl-5-thiazolesulfonyl chloride, 97%

Catalog No. AC148360010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC148360010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC148360010 Supplier Thermo Scientific Chemicals Supplier No. 148360010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 69812-29-9 | C6H7ClN2O3S2 | 254.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69812-29-9
Molecular Formula C6H7ClN2O3S2
Molecular Weight (g/mol) 254.71
MDL Number MFCD00005320
InChI Key NXGKPRKPUCSEIL-UHFFFAOYSA-N
Synonym 2-acetamido-4-methylthiazole-5-sulfonyl chloride, 2-acetamido-4-methyl-5-thiazolesulfonyl chloride, 2-acetylamino-4-methyl-thiazole-5-sulfonyl chloride, 5-thiazolesulfonyl chloride, 2-acetylamino-4-methyl, 2-acetamido-4-methylthiazole-5-sulphonyl chloride, 2-acetylamino-4-methyl-1,3-thiazole-5-sulfonyl chloride, 2-acetylamino-4-methylthiazole-5-sulfonyl chloride, 2-acetylamino-4-methyl-1,3-thiazol-5-ylsulphonyl chloride, n-5-chlorosulfonyl-4-methyl-1,3-thiazol-2-yl acetamide, ksc355a5n
PubChem CID 96951
IUPAC Name 2-acetamido-4-methyl-1,3-thiazole-5-sulfonyl chloride
SMILES CC1=C(SC(=N1)NC(=O)C)S(=O)(=O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 161°C to 164°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 254.71
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetamido-4-methyl-5-thiazolesulfonyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 274-127-7

RUO – Research Use Only