missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamido-3,4,6-tri-O-benzyl-2-deoxy-beta-D-glucopyranosyl Azide 98.0+%, TCI America™

Catalog No. A16785G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16785G 5 g
A16781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A16785G Supplier TCI America Supplier No. A16785G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 214467-60-4
Molecular Formula C29H32N4O5
Molecular Weight (g/mol) 516.598
MDL Number MFCD06797049
InChI Key XOOBJDMHDWWHLZ-HWVUQVAQSA-N
PubChem CID 15379132
IUPAC Name N-[(2R,3R,4R,5S,6R)-2-azido-4,5-bis(phenylmethoxy)-6-(phenylmethoxymethyl)oxan-3-yl]acetamide
SMILES CC(=O)NC1C(C(C(OC1N=[N+]=[N-])COCC2=CC=CC=C2)OCC3=CC=CC=C3)OCC4=CC=CC=C4

Specifications

Color White
Quantity 5 g
Formula Weight 516.60
Percent Purity ≥98.0% (HPLC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetamido-3,4,6-tri-O-benzyl-2-deoxy-beta-D-glucopyranosyl Azide
TSCA No
Recommended Storage Freezer