missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-alpha-D-glucopyranosyl Chloride 93.0+%, TCI America™

Catalog No. A14161G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14161G 1 g
A14165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14161G Supplier TCI America Supplier No. A14161G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3068-34-6
Molecular Formula C14H20ClNO8
Molecular Weight (g/mol) 365.763
MDL Number MFCD00069776
InChI Key NAYYKQAWUWXLPD-KSTCHIGDSA-N
Synonym 2-Acetamido-2-deoxy-alpha-D-glucopyranosyl Chloride 3,4,6-Triacetate, 1-Chloro-2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-alpha-D-glucose
PubChem CID 11100622
IUPAC Name [(2R,3S,4R,5R,6R)-5-acetamido-3,4-diacetyloxy-6-chlorooxan-2-yl]methyl acetate
SMILES CC(=O)NC1C(C(C(OC1Cl)COC(=O)C)OC(=O)C)OC(=O)C

Specifications

Quantity 1 g
Formula Weight 365.76
Percent Purity ≥93.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-alpha-D-glucopyranosyl Chloride
TSCA No
Recommended Storage Freezer