missing translation for 'onlineSavingsMsg'
Learn More

2-(9,9-Diphenyl-9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97.0+%, TCI America™

Catalog No. D50091G
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50091G 1 g
D5009200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50091G Supplier TCI America Supplier No. D50091G

Chemical Identifiers

CAS 462128-39-8
Molecular Formula C31H29BO2
Molecular Weight (g/mol) 444.38
MDL Number MFCD29089356
InChI Key CQMUDHYVDPNPLZ-UHFFFAOYSA-N
Synonym (9,9-Diphenyl-9H-fluoren-2-yl)boronic Acid Pinacol Ester, 9,9-Diphenyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-fluorene
PubChem CID 12014049
IUPAC Name 2-(9,9-diphenyl-9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC2=C(C=C1)C1=CC=CC=C1C2(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 200°C
Color White
Quantity 1 g
Formula Weight 444.38
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(9,9-Diphenyl-9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane