missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dihydroxynaphthalene 99.0+%, TCI America™

Catalog No. D059425G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D059425G 25 g
D0594100G 100 g
D0594500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D059425G Supplier TCI America Supplier No. D059425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 582-17-2
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.17
MDL Number MFCD00004085
InChI Key DFQICHCWIIJABH-UHFFFAOYSA-N
Synonym 2,7-dihydroxynaphthalene, 2,7-naphthalenediol, naphthalenediol-2,7, unii-0to8e448ud, naphthalenediol-2,7 french, 2,7-dihydroxy naphthalene, 2,7-naphthalene diol, naphthalene-2-7-diol, 2,7-dihdroxynaphthalene
PubChem CID 11397
IUPAC Name naphthalene-2,7-diol
SMILES OC1=CC2=CC(O)=CC=C2C=C1

Specifications

Melting Point 186°C
Quantity 25 g
Formula Weight 160.17
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dihydroxynaphthalene
EINECSNumber (4)-0356
RTECSNumber QJ4780000
TSCA Yes