missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dibromotriphenylene 98.0+%, TCI America™

Catalog No. D4801200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4801200MG 200 mg
D48011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4801200MG Supplier TCI America Supplier No. D4801200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 888041-37-0
Molecular Formula C18H10Br2
Molecular Weight (g/mol) 386.086
MDL Number MFCD22571695
InChI Key BPGPBYGXGRDFQG-UHFFFAOYSA-N
PubChem CID 16122224
IUPAC Name 2,7-dibromotriphenylene
SMILES C1=CC=C2C(=C1)C3=C(C=CC(=C3)Br)C4=C2C=C(C=C4)Br

Specifications

Melting Point 232°C
Color Yellow
Quantity 200 mg
Formula Weight 386.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dibromotriphenylene