missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dibromo-9-fluorenone, 96%

Catalog No. AAH2591903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2591903 1 g
AAH2591906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2591903 Supplier Thermo Scientific Chemicals Supplier No. H2591903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 14348-75-5 | C13H6Br2O | 337.998 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14348-75-5
Molecular Formula C13H6Br2O
Molecular Weight (g/mol) 337.998
MDL Number MFCD00010790
InChI Key CWGRCRZFJOXQFV-UHFFFAOYSA-N
Synonym 2,7-dibromo-9-fluorenone, 2,7-dibromo-9h-fluoren-9-one, 2,7-dibromo-fluoren-9-one, 2,7-dibromofluorenone, 2,7-dibromo-fluorenone, 9h-fluoren-9-one, 2,7-dibromo, 2,7-dibrom-9h-fluoren-9-on, pubchem19652, acmc-209cqp, maybridge1_002427
PubChem CID 259922
IUPAC Name 2,7-dibromofluoren-9-one
SMILES C1=CC2=C(C=C1Br)C(=O)C3=C2C=CC(=C3)Br

Specifications

Melting Point 195°C to 196°C
Quantity 1 g
Formula Weight 337.99
Percent Purity 96%
Chemical Name or Material 2,7-Dibromo-9-fluorenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only