missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dibromo-9,9-dimethylfluorene 98.0+%, TCI America™

Catalog No. D38595G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38595G 5 g
D385925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38595G Supplier TCI America Supplier No. D38595G

Chemical Identifiers

CAS 28320-32-3
Molecular Formula C15H12Br2
Molecular Weight (g/mol) 352.07
MDL Number MFCD00099471
InChI Key LONBOJIXBFUBKQ-UHFFFAOYSA-N
Synonym 2,7-dibromo-9,9-dimethyl-9h-fluorene, 9,9-dimethyl-2,7-dibromofluorene, 2,7-dibromo-9,9-dimethyl-fluorene, 9h-fluorene, 2,7-dibromo-9,9-dimethyl, 2,7-dibromo-9,9-dimethyl fluorene, pubchem19656, acmc-1cpi8, ksc491i1n, 9,9-dimethyl-2,7-dibromofluorenone
PubChem CID 12123876
IUPAC Name 2,7-dibromo-9,9-dimethyl-9H-fluorene
SMILES CC1(C)C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2

Specifications

Melting Point 179°C
Quantity 5 g
Formula Weight 352.07
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dibromo-9,9-dimethylfluorene