missing translation for 'onlineSavingsMsg'
Learn More

2,6-Pyridinedicarboxaldehyde, 97%

Catalog No. AC437752500
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC437752500 250 mg Glass bottle
AC437750010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC437752500 Supplier Thermo Scientific Chemicals Supplier No. 437752500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5431-44-7 | C7H5NO2 | 135.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5431-44-7
Molecular Formula C7H5NO2
Molecular Weight (g/mol) 135.12
InChI Key PMWXGSWIOOVHEQ-UHFFFAOYSA-N
Synonym 2,6-pyridinedicarboxaldehyde, 2,6-diformylpyridine, 2,6-pyridine dialdehyde, 2,6-pyridinedicarbaldehyde, pubchem22285, acmc-1ap0d, pyda72, 2,6-pyridinedicarboxadehyde, 2,6-pyridine dicarboxaldehyde, 2,6-pyridinedicarbaldehyde #
PubChem CID 79485
IUPAC Name pyridine-2,6-dicarbaldehyde
SMILES C1=CC(=NC(=C1)C=O)C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 123.3°C to 124.5°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 250 mg
Formula Weight 135.12
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2, 6-Pyridinedicarboxaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 226-589-6

RUO – Research Use Only