missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dinitroaniline 95.0+%, TCI America™

Catalog No. D081510G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D081510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D081510G Supplier TCI America Supplier No. D081510G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 606-22-4
Molecular Formula C6H5N3O4
Molecular Weight (g/mol) 183.123
MDL Number MFCD00007148
InChI Key QFUSCYRJMXLNRB-UHFFFAOYSA-N
Synonym benzenamine, 2,6-dinitro, 2,6-dinitrobenzenamine, aniline, 2,6-dinitro, unii-8w27g0qbd7, ccris 3108, pubchem22911, wln: wnr bz cnw, acmc-209ml1, 2,6-dinitroaniline, 2,6-dinitro-phenyl-amine
PubChem CID 69070
IUPAC Name 2,6-dinitroaniline
SMILES C1=CC(=C(C(=C1)[N+](=O)[O-])N)[N+](=O)[O-]

Specifications

Melting Point 138°C
Quantity 10 g
UN Number 1596
Formula Weight 183.12
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dinitroaniline
RTECSNumber BX9200000
TSCA No