missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dinitro-4-(trifluoromethyl)phenol 97.0+%, TCI America™
Supplier: TCI America D28805G
Specifications
| 2,6-Dinitro-4-(trifluoromethyl)phenol | |
| Yellow | |
| MFCD00194260 | |
| 2930 | |
| FXZGYEWQIGIFMC-UHFFFAOYSA-N | |
| 2,6-dinitro-4-(trifluoromethyl)phenol | |
| 164591 | |
| ≥97.0% (GC,T) |
| 393-77-1 | |
| C7H3F3N2O5 | |
| 5 g | |
| 4-Hydroxy-3,5-dinitrobenzotrifluoride, alpha,alpha,alpha-Trifluoro-2,6-dinitro-p-cresol | |
| C1=C(C=C(C(=C1[N+](=O)[O-])O)[N+](=O)[O-])C(F)(F)F | |
| 252.105 | |
| 252.11 | |
| Crystalline Powder |
Chemical Identifiers
| 393-77-1 | |
| 252.105 | |
| FXZGYEWQIGIFMC-UHFFFAOYSA-N | |
| 164591 | |
| C1=C(C=C(C(=C1[N+](=O)[O-])O)[N+](=O)[O-])C(F)(F)F |
| C7H3F3N2O5 | |
| MFCD00194260 | |
| 4-Hydroxy-3,5-dinitrobenzotrifluoride, alpha,alpha,alpha-Trifluoro-2,6-dinitro-p-cresol | |
| 2,6-dinitro-4-(trifluoromethyl)phenol |
Safety and Handling
TSCA : No