missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dinitro-4-(trifluoromethyl)phenol 97.0+%, TCI America™

Catalog No. p-7127553
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28805G 5 g
D288025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28805G Supplier TCI America Supplier No. D28805G
Only null left

Chemical Identifiers

CAS 393-77-1
Molecular Formula C7H3F3N2O5
Molecular Weight (g/mol) 252.105
MDL Number MFCD00194260
InChI Key FXZGYEWQIGIFMC-UHFFFAOYSA-N
Synonym 4-Hydroxy-3,5-dinitrobenzotrifluoride, alpha,alpha,alpha-Trifluoro-2,6-dinitro-p-cresol
PubChem CID 164591
IUPAC Name 2,6-dinitro-4-(trifluoromethyl)phenol
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])O)[N+](=O)[O-])C(F)(F)F

Specifications

Color Yellow
Quantity 5 g
UN Number 2930
Formula Weight 252.11
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dinitro-4-(trifluoromethyl)phenol