missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethyl-4-nitrophenol, 98%

Catalog No. AAA1547214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1547214 25 g
AAA1547206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1547214 Supplier Thermo Scientific Chemicals Supplier No. A1547214

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2423-71-4 | C8H9NO3 | 167.164 g/mol

It was used as internal standard in determination of 4-nitrophenol and 3-methyl-4-nitrophenol in human urine by liquid chromatography combined with tandem mass spectrometry. ,6-Dimethyl-4-nitrophenol was used to study the disposition requirement for binding of p-nitrophenol and sodium p-nitrophenolate like substrates with cyclohexaamylaose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It was used as internal standard in determination of 4-nitrophenol and 3-methyl-4-nitrophenol in human urine by liquid chromatography combined with tandem mass spectrometry. ,6-Dimethyl-4-nitrophenol was used to study the disposition requirement for binding of p-nitrophenol and sodium p-nitrophenolate like substrates with cyclohexaamylaose.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 2423-71-4
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00007339
InChI Key FNORUNUDZNWQFF-UHFFFAOYSA-N
PubChem CID 17030
IUPAC Name 2,6-dimethyl-4-nitrophenol
SMILES CC1=CC(=CC(=C1O)C)[N+](=O)[O-]

Specifications

Melting Point ∼168°C (decomposition)
Quantity 25 g
Beilstein 1873275
Synonym 4-nitro-2,6-xylenol, 2,6-xylenol, 4-nitro, 4-nitro-2,6-dimethylphenol, phenol, 2,6-dimethyl-4-nitro, phenol,2,6-dimethyl-4-nitro, acmc-1cljn, 2, 4-nitro, intermediates-zcf02651, 2,6-dimethyl4-nitrophenol, phenol,6-dimethyl-4-nitro
Solubility Information Insoluble in water.
Formula Weight 167.17
Percent Purity 98%
Chemical Name or Material 2,6-Dimethyl-4-nitrophenol
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
EINECSNumber 219-353-9
RTECSNumber ZE8225000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only