missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethyl-4-hydroxypyridine 98.0+%, TCI America™

Catalog No. D36381G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36381G 1 g
D36385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36381G Supplier TCI America Supplier No. D36381G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13603-44-6
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00956021
InChI Key PRAFLUMTYHBEHE-UHFFFAOYSA-N
Synonym 2,6-Dimethyl-4-pyridinol
PubChem CID 83609
IUPAC Name 2,6-dimethyl-1H-pyridin-4-one
SMILES CC1=CC(=O)C=C(N1)C

Specifications

Melting Point 230°C
Boiling Point 351°C
Quantity 1 g
Formula Weight 123.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dimethyl-4-hydroxypyridine