missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethyl-4-heptanone 90.0+%, TCI America™

Catalog No. D0733500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0733500ML 500 mL
D073325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0733500ML Supplier TCI America Supplier No. D0733500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-83-8
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
MDL Number MFCD00008940
InChI Key PTTPXKJBFFKCEK-UHFFFAOYSA-N
Synonym 2,6-dimethyl-4-heptanone, diisobutyl ketone, isovalerone, isobutyl ketone, valerone, diisobutylketone, diisobutilchetone, s-diisopropylacetone, di-isobutylcetone, dibk
PubChem CID 7958
IUPAC Name 2,6-dimethylheptan-4-one
SMILES CC(C)CC(=O)CC(C)C

Specifications

Color Yellow
Boiling Point 168°C
Quantity 500 mL
UN Number 1157
Formula Weight 142.24
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,6-Dimethyl-4-heptanone
EINECSNumber (2)-2475
RTECSNumber MJ5775000
TSCA Yes