missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethyl-3,5-heptanedione 97.0+%, TCI America™

Catalog No. D24135G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24135G 5 g
D241325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24135G Supplier TCI America Supplier No. D24135G

Chemical Identifiers

CAS 18362-64-6
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.225
MDL Number MFCD00015040
InChI Key CEGGECULKVTYMM-UHFFFAOYSA-N
Synonym 2,6-dimethyl-3,5-heptanedione, diisobutyrylmethane, 3,5-heptanedione, 2,6-dimethyl, acmc-209ekw, 2,6-dimethyl-3,5-heptandione
PubChem CID 87597
IUPAC Name 2,6-dimethylheptane-3,5-dione
SMILES CC(C)C(=O)CC(=O)C(C)C

Specifications

Color Yellow
Boiling Point 80°C
Quantity 5 g
UN Number 1224
Formula Weight 156.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,6-Dimethyl-3,5-heptanedione