missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethoxypyridine-3-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. D46901G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D46901G Supplier TCI America Supplier No. D46901G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 2,6-Dimethoxypyridine-3-boronic Acid (contains varying amounts of Anhydride)
CAS 221006-70-8
Melting Point 109°C
Color White
Molecular Formula C7H10BNO4
Quantity 1 g
MDL Number MFCD03788239
Synonym 2,6-dimethoxypyridine-3-boronic acid, 2,6-dimethoxy-3-pyridineboronic acid, 2,6-dimethoxypyridin-3-yl boronic acid, 2,6-dimethoxypyridin-3-yl-3-boronic acid, 3-borono-2,6-dimethoxypyridine, 2,6-dimethoxy-3-pyridylboronic acid, 2,6-dimethoxypyridine-5-boronic acid, boronic acid, 2,6-dimethoxy-3-pyridinyl, 2,6-dimethoxy-3-pyridyl boronic acid
InChI Key ADGHSWFUZUADDH-UHFFFAOYSA-N
SMILES B(C1=C(N=C(C=C1)OC)OC)(O)O
IUPAC Name (2,6-dimethoxypyridin-3-yl)boronic acid
Molecular Weight (g/mol) 182.97
PubChem CID 2762707
Formula Weight 182.97
Physical Form Crystalline Powder
Show More Show Less