missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzenesulfonyl chloride, 97%

Catalog No. AAA1359503
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1359503 1 g
AAA1359506 5 g
AAA1359514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1359503 Supplier Thermo Scientific Chemicals Supplier No. A1359503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 60230-36-6 | C6H3ClF2O2S | 212.595 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 60230-36-6
Molecular Formula C6H3ClF2O2S
Molecular Weight (g/mol) 212.595
MDL Number MFCD00085004
InChI Key QXWAUQMMMIMLTO-UHFFFAOYSA-N
Synonym 2,6-difluorobenzene-1-sulfonyl chloride, 2,6-difluorobenzenesulfonylchloride, 2,6-difluorobenzenesulphonyl chloride, benzenesulfonyl chloride, 2,6-difluoro, 2,6-difluoro-benzenesulfonyl chloride, 2,6-difluorobenzene sulfonyl chloride, 2,6-difluorophenyl chlorosulfone, pubchem1957, acmc-209mil, ksc352q0t
PubChem CID 2734271
IUPAC Name 2,6-difluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C(=C1)F)S(=O)(=O)Cl)F

Specifications

Density 1.568
Boiling Point 127°C to 130°C (6.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.526
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 212.6
Percent Purity 97%
Chemical Name or Material 2,6-Difluorobenzenesulfonyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only