missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoro-3-hydroxybenzaldehyde, 95%

Catalog No. AAH32250MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32250MD 250 mg
AAH3225003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH32250MD Supplier Thermo Scientific Chemicals Supplier No. H32250MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 152434-88-3 | C7H4F2O2 | 158.104 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 152434-88-3
Molecular Formula C7H4F2O2
Molecular Weight (g/mol) 158.104
MDL Number MFCD08275284
InChI Key NHGSYGIEJAONJB-UHFFFAOYSA-N
PubChem CID 594111
IUPAC Name 2,6-difluoro-3-hydroxybenzaldehyde
SMILES C1=CC(=C(C(=C1O)F)C=O)F

Specifications

Melting Point 110°C to 114°C
Quantity 250 mg
Sensitivity Air Sensitive
Synonym 2,6-difluoro-3-hydroxy-benzaldehyde, benzaldehyde,2,6-difluoro-3-hydroxy, acmc-1c55f, 2,4-difluoro-3-formylphenol, #, benzaldehyde, 2,6-difluoro-3-hydroxy, 2,6-bis fluoranyl-3-oxidanyl-benzaldehyde, benzaldehyde, 2,6-difluoro-3-hydroxy-9ci
Formula Weight 158.11
Percent Purity 95%
Chemical Name or Material 2,6-Difluoro-3-hydroxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only