missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenylacetic Acid 96.0+%, TCI America™

Catalog No. D189725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D189725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D189725G Supplier TCI America Supplier No. D189725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6575-24-2
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.034
MDL Number MFCD00004320
InChI Key SFAILOOQFZNOAU-UHFFFAOYSA-N
Synonym 2,6-dichlorophenylacetic acid, 2-2,6-dichlorophenyl acetic acid, 2,6-dichlorophenyl acetic acid, benzeneacetic acid, 2,6-dichloro, 2,6-dichloro benzeneacetic acid, 2,6-dichlorophenylaceticacid, unii-k8g025cx8w, 2,6-dichlorobenzeneaceticacid, 2cl
PubChem CID 81058
IUPAC Name 2-(2,6-dichlorophenyl)acetic acid
SMILES C1=CC(=C(C(=C1)Cl)CC(=O)O)Cl

Specifications

Melting Point 160°C
Quantity 25 g
Formula Weight 205.03
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichlorophenylacetic Acid